4-(trans-4-Butylcyclohexyl)phenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5914-1GEA 291 EUR
COBBBB-5914-1G
4-(trans-4-Butylcyclohexyl)phenylboronsäure
4-(trans-4-Butylcyclohexyl)phenylboronsäure
Formel: C₁₆H₂₅BO₂
MDL: MFCD09750934
CAS-Nummer: 516510-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR