2-(Chlormethyl)-5-nitrobenzoxazol

Lieferant: SIA ENAMINE

ENAMEN300102161.1GEA 200 EUR
ENAMEN300102161.1G ENAMEN300102161.5G ENAMEN300102161
2-(Chlormethyl)-5-nitrobenzoxazol
2-(Chloromethyl)-5-nitrobenzoxazole
Formel: C₈H₅ClN₂O₃
MDL: MFCD01596292
CAS-Nummer: 119198-10-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR