2-Amino-N-(4-methylphenyl)benzamid

Lieferant: SIA ENAMINE

ENAMEN30006124.1GEA 0 EUR
ENAMEN30006124 ENAMEN30006124.10G ENAMEN30006124.5G ENAMEN30006124.1G
2-Amino-N-(4-methylphenyl)benzamid
2-Amino-N-(4-methylphenyl)benzamid
Formel: C₁₄H₁₄N₂O
Molecular Weight: 226,2700 g/mol
MDL: MFCD00115975
CAS-Nummer: 32212-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR