4-Fluor-2-methylbenzolsulfonamid

Lieferant: SIA ENAMINE

ENAMEN30001207.1GEA 0 EUR
ENAMEN30001207.5G ENAMEN30001207.10G ENAMEN30001207.1G
4-Fluor-2-methylbenzolsulfonamid
4-Fluor-2-methylbenzolsulfonamid
Formel: C₇H₈FNO₂S
Molecular Weight: 189,2073 g/mol
MDL: MFCD02709831
CAS-Nummer: 489-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR