5-(2-Methylphenyl)-1H-tetrazol

Lieferant: SIA ENAMINE

ENAMEN30002918.1GEA 0 EUR
ENAMEN30002918.5G ENAMEN30002918.10G ENAMEN30002918.1G
5-(2-Methylphenyl)-1H-tetrazol
5-(2-Methylphenyl)-1H-tetrazole
Formel: C₈H₈N₄
Molecular Weight: 160,18 g/mol
MDL: MFCD03093095
CAS-Nummer: 51449-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR