3-Amino-2,5-dibrompyridin

Lieferant: SIA ENAMINE

ENAMEN300107027.1GEA 0 EUR
ENAMEN300107027 ENAMEN300107027.1G ENAMEN300107027.5G
3-Amino-2,5-dibrompyridin
3-Amino-2,5-dibromopyridine
Formel: C₅H₄Br₂N₂
Molecular Weight: 251,9100 g/mol
MDL: MFCD09266224
CAS-Nummer: 90902-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR