5-(Chlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol

Lieferant: SIA ENAMINE

ENAMEN30010545.1GEA 38.2 EUR
ENAMEN30010545.1G ENAMEN30010545.10G ENAMEN30010545.5G
5-(Chlormethyl)-3-(4-methylphenyl)-1,2,4-oxadiazol
5-(Chloromethyl)-3-(4-methylphenyl)-1,2,4-oxadiazole
Formel: C₁₀H₉ClN₂O
MDL: MFCD06093407
CAS-Nummer: 50737-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR