4-Amino-N,N-dimethylbenzamid

Lieferant: SIA ENAMINE

ENAMEN30009518.1GEA 0 EUR
ENAMEN30009518.5G ENAMEN30009518.10G ENAMEN30009518.1G
4-Amino-N,N-dimethylbenzamid
4-Amino-N,N-dimethylbenzamid
Formel: C₉H₁₂N₂O
MDL: MFCD00523648
CAS-Nummer: 6331-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR