2-Amino-4-methoxybenzonitril

Lieferant: SIA ENAMINE

ENAMEN300106785.1GEA 0 EUR
ENAMEN300106785.1G ENAMEN300106785.5G ENAMEN300106785
2-Amino-4-methoxybenzonitril
2-Amino-4-methoxybenzonitril
Formel: C₈H₈N₂O
Molecular Weight: 148,1600 g/mol
MDL: MFCD05149246
CAS-Nummer: 38487-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR