4-Brom-2,5-difluorthiophenol

Lieferant: SIA ENAMINE

ENAMEN300102765.1GEA 0 EUR
ENAMEN300102765.1G ENAMEN300102765 ENAMEN300102765.5G
4-Brom-2,5-difluorthiophenol
4-Brom-2,5-difluorthiophenol
Formel: C₆H₃BrF₂S
MDL: MFCD12026144
CAS-Nummer: 1208075-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR