5-Amino-3-(4-chlorphenyl)isoxazol
Lieferant: SIA ENAMINE
ENAMEN30009527.1GEA
0
EUR
ENAMEN30009527.5G
ENAMEN30009527.10G
ENAMEN30009527.1G
5-Amino-3-(4-chlorphenyl)isoxazol
5-Amino-3-(4-chlorophenyl)isoxazole
Formel:
C₉H₇ClN₂O Molecular Weight: 194,6177 g/mol |
MDL:
MFCD01569450 CAS-Nummer: 33866-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...