3,4-(Ethylendioxy)thiophenol

Lieferant: SIA ENAMINE

ENAMEN30005854.1GEA 0 EUR
ENAMEN30005854.1G ENAMEN30005854.5G ENAMEN30005854 ENAMEN30005854.10G
3,4-(Ethylendioxy)thiophenol
3,4-(Ethylendioxy)thiophenol
Formel: C₈H₈O₂S
MDL: MFCD06201753
CAS-Nummer: 247228-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR