3-Amino-5-(trifluormethyl)-1H-indazol

Lieferant: SIA ENAMINE

ENAMEN300100296.1GEA 0 EUR
ENAMEN300100296.1G ENAMEN300100296.5G ENAMEN300100296
3-Amino-5-(trifluormethyl)-1H-indazol
3-Amino-5-(trifluoromethyl)-1H-indazole
Formel: C₈H₆F₃N₃
MDL: MFCD01719740
CAS-Nummer: 2250-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR