Methyl-2,3,4-trifluorbenzoat

Lieferant: SIA ENAMINE

ENAMEN300101993.1GEA 0 EUR
ENAMEN300101993.1G ENAMEN300101993.5G ENAMEN300101993
Methyl-2,3,4-trifluorbenzoat
Methyl-2,3,4-trifluorbenzoat
Formel: C₈H₅F₃O₂
MDL: MFCD06201866
CAS-Nummer: 773873-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR