3-Chlor-6-(2-methyl-1-imidazolyl)pyridazin

Lieferant: SIA ENAMINE

ENAMEN300104274.1GEA 0 EUR
ENAMEN300104274.5G ENAMEN300104274 ENAMEN300104274.1G
3-Chlor-6-(2-methyl-1-imidazolyl)pyridazin
3-Chloro-6-(2-methyl-1-imidazolyl)pyridazine
Formel: C₈H₇ClN₄
MDL: MFCD09879090
CAS-Nummer: 75792-66-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR