4-Fluor-3-nitrobenzolsulfonylchlorid

Lieferant: SIA ENAMINE

ENAMEN30010260.1GEA 0 EUR
ENAMEN30010260.10G ENAMEN30010260.1G ENAMEN30010260 ENAMEN30010260.5G
4-Fluor-3-nitrobenzolsulfonylchlorid
4-Fluor-3-nitrobenzolsulfonylchlorid
Formel: C₆H₃ClFNO₄S
Molecular Weight: 239,6100 g/mol
MDL: MFCD06342770
CAS-Nummer: 6668-56-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR