1-(4-Chlorphenylsulfonyl)piperazin

Lieferant: SIA ENAMINE

ENAMEN30004728.1GEA 0 EUR
ENAMEN30004728.10G ENAMEN30004728.1G ENAMEN30004728.5G
1-(4-Chlorphenylsulfonyl)piperazin
1-(4-Chlorphenylsulfonyl)piperazin
Formel: C₁₀H₁₃ClN₂O₂S
MDL: MFCD00973893
CAS-Nummer: 16017-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR