4-(Benzyloxy)-3-brombenzaldehyd

Lieferant: SIA ENAMINE

ENAMEN30005378.1GEA 0 EUR
ENAMEN30005378.1G ENAMEN30005378.10G ENAMEN30005378.5G
4-(Benzyloxy)-3-brombenzaldehyd
4-(Benzyloxy)-3-brombenzaldehyd
Formel: C₁₄H₁₁BrO₂
Molecular Weight: 291,1400 g/mol
CAS-Nummer: 69455-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR