3',4'-Dimethyl-2-phenylacetophenon

Lieferant: SIA ENAMINE

ENAMEN300101848.1GEA 677 EUR
ENAMEN300101848.1G ENAMEN300101848.5G ENAMEN300101848
3',4'-Dimethyl-2-phenylacetophenon
3',4'-Dimethyl-2-phenylacetophenon
Formel: C₁₆H₁₆O
MDL: MFCD00156722
CAS-Nummer: 65614-75-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR