5'-Brom-3'-chlor-2'-hydroxyacetophenon

Lieferant: SIA ENAMINE

ENAMEN300106332.1GEA 0 EUR
ENAMEN300106332.5G ENAMEN300106332.1G ENAMEN300106332
5'-Brom-3'-chlor-2'-hydroxyacetophenon
5'-Brom-3'-chlor-2'-hydroxyacetophenon
Formel: C₈H₆BrClO₂
MDL: MFCD02660600
CAS-Nummer: 331821-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR