4-Fluor-N-methylbenzylamin

Lieferant: SIA ENAMINE

ENAMEN30007965.1GEA 0 EUR
ENAMEN30007965.5G ENAMEN30007965.1G ENAMEN30007965.10G
4-Fluor-N-methylbenzylamin
4-Fluor-N-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD03839848
CAS-Nummer: 405-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR