ω-Chlor-3,4-difluoracetophenon

Lieferant: SIA ENAMINE

ENAMEN30001950.1GEA 0 EUR
ENAMEN30001950.5G ENAMEN30001950.1G ENAMEN30001950 ENAMEN30001950.10G
ω-Chlor-3,4-difluoracetophenon
ω-Chlor-3,4-difluoracetophenon
Formel: C₈H₅ClF₂O
Molecular Weight: 190,5745 g/mol
MDL: MFCD03966888
CAS-Nummer: 51336-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR