4-Amino-α,α,α-trifluor-o-toluylsäure

Lieferant: SIA ENAMINE

ENAMEN300105577.1GEA 0 EUR
ENAMEN300105577.1G ENAMEN300105577.5G ENAMEN300105577
4-Amino-α,α,α-trifluor-o-toluylsäure
4-Amino-α,α,α-trifluor-o-toluylsäure
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD03407959
CAS-Nummer: 393-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR