5-Nitro-2-thiophencarbonsäure

Lieferant: SIA ENAMINE

ENAMEN30000713.1GEA 0 EUR
ENAMEN30000713.10G ENAMEN30000713.5G ENAMEN30000713.1G ENAMEN30000713
5-Nitro-2-thiophencarbonsäure
5-Nitro-2-thiophenecarboxylic acid
Formel: C₅H₃NO₄S
Molecular Weight: 173,14 g/mol
MDL: MFCD00159552
CAS-Nummer: 6317-37-9
EINECS: 228-654-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR