2-Amino-4-(4-chlorphenyl)thiazol
Lieferant: SIA ENAMINE
ENAMEN30002225.1GEA
0
EUR
ENAMEN30002225.1G
ENAMEN30002225.10G
ENAMEN30002225.5G
2-Amino-4-(4-chlorphenyl)thiazol
2-Amino-4-(4-chlorophenyl)thiazole
Formel:
C₉H₇ClN₂S Molecular Weight: 210,69 g/mol |
MDL:
MFCD00047058 CAS-Nummer: 2103-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...