Methyl-3-formylbenzoat

Lieferant: SIA ENAMINE

ENAMEN300106923.1GEA 0 EUR
ENAMEN300106923.5G ENAMEN300106923.1G ENAMEN300106923
Methyl-3-formylbenzoat
Methyl-3-formylbenzoat
Formel: C₉H₈O₃
MDL: MFCD00189379
CAS-Nummer: 52178-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR