3-(3-Fluor-4-methoxybenzoyl)propionsäure
Lieferant: SIA ENAMINE
ENAMEN300101432.1GEA
0
EUR
ENAMEN300101432.1G
ENAMEN300101432.5G
ENAMEN300101432
3-(3-Fluor-4-methoxybenzoyl)propionsäure
3-(3-Fluor-4-methoxybenzoyl)propionsäure
Formel:
C₁₁H₁₁FO₄ Molecular Weight: 226,20 g/mol |
MDL:
MFCD00020538 CAS-Nummer: 347-63-7 EINECS: 206-474-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...