4-Anilino-1-benzylpiperidin

Lieferant: SIA ENAMINE

ENAMEN30002588.1GEA 38.2 EUR
ENAMEN30002588.10G ENAMEN30002588.5G ENAMEN30002588 ENAMEN30002588.1G
4-Anilino-1-benzylpiperidin
4-Anilino-1-benzylpiperidin
Formel: C₁₈H₂₂N₂
MDL: MFCD00023738
CAS-Nummer: 1155-56-2
EINECS: 214-583-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR