2-Amino-4-(3-pyridyl)thiazol

Lieferant: SIA ENAMINE

Synonyme: 4-(Pyridin-3-yl)thiazol-2-amine

EN30002252.1G
ENAMEN30002252.1GEA 0 EUR
ENAMEN30002252.1G
2-Amino-4-(3-pyridyl)thiazol
2-Amino-4-(3-pyridyl)thiazole
Formel: C₈H₇N₃S
Molecular Weight: 177,22 g/mol
MDL: MFCD00218013
CAS-Nummer: 30235-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR