2-Amino-4-(2-pyridyl)thiazol

Lieferant: SIA ENAMINE

ENAMEN30002253.1GEA 0 EUR
ENAMEN30002253 ENAMEN30002253.5G ENAMEN30002253.10G ENAMEN30002253.1G
2-Amino-4-(2-pyridyl)thiazol
2-Amino-4-(2-pyridyl)thiazole
Formel: C₈H₇N₃S
Molecular Weight: 177,22 g/mol
MDL: MFCD00460417
CAS-Nummer: 30235-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR