2-Methyl-5-pyridinboronsäure Hydrat

Lieferant: COMBI-BLOCKS INC.

COBBBB-8495-1GEA 194 EUR
COBBBB-8495-1G COBBBB-8495-5G
2-Methyl-5-pyridinboronsäure Hydrat
2-Methyl-5-pyridinboronsäure Hydrat
Formel: C₆H₈BNO₂·H₂O
MDL: MFCD06657901
CAS-Nummer: 1072952-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR