Methyl-4-brom-3-(trifluormethyl)benzoat

Lieferant: COMBI-BLOCKS INC.

COBBQA-0897-5GEA 109 EUR
COBBQA-0897-5G
Methyl-4-brom-3-(trifluormethyl)benzoat
Methyl-4-brom-3-(trifluormethyl)benzoat
Formel: C₉H₆BrF₃O₂
Molecular Weight: 283,0420 g/mol
MDL: MFCD10566409
CAS-Nummer: 107317-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR