3-(2-Methylphenyl)-5-phenyl-1,2,4-oxadiazol ≥97%

Achtung

H54927.MDEA 32.34 EUR
H54927.MD
3-(2-Methylphenyl)-5-phenyl-1,2,4-oxadiazol ≥97%
3-(2-Methylphenyl)-5-phenyl-1,2,4-oxadiazole
Formel: C₁₅H₁₂N₂O
Molecular Weight: 236,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20265377
CAS-Nummer: 1400191-58-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR