3-(4-Bromphenyl)-5-(dichlormethyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6566-5GEA 175 EUR
COBBHC-6566-5G COBBHC-6566-1G
3-(4-Bromphenyl)-5-(dichlormethyl)-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-(dichloromethyl)-1,2,4-oxadiazole
Formel: C₉H₅BrCl₂N₂O
MDL: MFCD11855937
CAS-Nummer: 1133116-19-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR