3-(4-Bromphenyl)-5-butyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6437-5GEA 329 EUR
COBBHC-6437-5G COBBHC-6437-1G
3-(4-Bromphenyl)-5-butyl-1,2,4-oxadiazol
3-(4-Bromophenyl)-5-butyl-1,2,4-oxadiazole
Formel: C₁₂H₁₃BrN₂O
MDL: MFCD00635531
CAS-Nummer: 313536-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR