3-(3-Bromphenyl)-5-propyl-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-2517-1GEA 72 EUR
COBBHC-2517-1G COBBHC-2517-5G
3-(3-Bromphenyl)-5-propyl-1,2,4-oxadiazol
3-(3-Bromophenyl)-5-propyl-1,2,4-oxadiazole
Formel: C₁₁H₁₁BrN₂O
MDL: MFCD12913950
CAS-Nummer: 1187385-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR