5-Benzyl-3-(2-bromphenyl)-1,2,4-oxadiazol

Lieferant: COMBI-BLOCKS INC.

COBBHC-6290-1GEA 135 EUR
COBBHC-6290-1G COBBHC-6290-5G
5-Benzyl-3-(2-bromphenyl)-1,2,4-oxadiazol
5-Benzyl-3-(2-bromphenyl)-1,2,4-oxadiazol
Formel: C₁₅H₁₁BrN₂O
MDL: MFCD10699653
CAS-Nummer: 1033201-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR