Methyl-2-iodisonicotinat

Lieferant: COMBI-BLOCKS INC.

COBBPY-1496-1GEA 290 EUR
COBBPY-1496-1G
Methyl-2-iodisonicotinat
Methyl-2-iodisonicotinat
Formel: C₇H₆INO₂
Molecular Weight: 263,0325 g/mol
MDL: MFCD08235123
CAS-Nummer: 134579-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR