Calciumchlorid Hydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ quality level = MQ100

SIAL202940-10GEA 208 EUR
SIAL202940-10G
Calciumchlorid Hydrat, Sigma-Aldrich®
Calciumchlorid Hydrat
Formel: CaCl₂·H₂O
Molecular Weight: 110,98 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149613
CAS-Nummer: 22691-02-7
EINECS: 233-140-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR