Methyl-4-brom-2-(brommethyl)benzoat

Lieferant: COMBI-BLOCKS INC.

COBBSH-5535-1GEA 116 EUR
COBBSH-5535-1G
Methyl-4-brom-2-(brommethyl)benzoat
Methyl-4-brom-2-(brommethyl)benzoat
Formel: C₉H₈Br₂O₂
Molecular Weight: 307,97 g/mol
MDL: MFCD04114319
CAS-Nummer: 78471-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR