2-Ethylbenzoesäure

Lieferant: COMBI-BLOCKS INC.

COBBSS-4809-1GEA 22 EUR
COBBSS-4809-1G
2-Ethylbenzoesäure
2-Ethylbenzoic acid
Formel: C₉H₁₀O₂
MDL: MFCD00045838
CAS-Nummer: 612-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR