5-Brom-o-toluylsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL694797-5GEA 58.4 EUR
SIAL694797-5G SIAL694797-1G
5-Brom-o-toluylsäure, Sigma-Aldrich®
5-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00267350
CAS-Nummer: 79669-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR