6-Chlor-2,3-difluorbenzaldehyd ≥97%

Achtung

H34287.03EA 28.4 EUR
H34287.03 H34287.06
6-Chlor-2,3-difluorbenzaldehyd ≥97%
6-Chlor-2,3-difluorbenzaldehyd
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368118
CAS-Nummer: 797791-33-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR