4-tert-Butoxybenzonitril 96%

Lieferant: ABCR GMBH
Achtung

ABCRAB180471.0025EA 0 EUR
ABCRAB180471.0025
4-tert-Butoxybenzonitril 96%
4-tert-Butoxybenzonitril
Formel: C₁₁H₁₃NO
Molecular Weight: 175,23 g/mol
Siedepunkt: 95 °C (0,8 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD03425273
CAS-Nummer: 185259-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR