3-Phenoxypropionitril

Lieferant: CHEMPUR

CHMPFL12260.POREA 0 EUR
CHMPFL12260.POR
3-Phenoxypropionitril
3-Phenoxypropionitril
Formel: C₉H₉NO
MDL: MFCD00013822
CAS-Nummer: 3055-86-5
EINECS: 221-278-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR