1-Brom-2,4-dichlor-5-nitrobenzol

Lieferant: COMBI-BLOCKS INC.

COBBAN-2607-5GEA 282 EUR
COBBAN-2607-5G
1-Brom-2,4-dichlor-5-nitrobenzol
1-Bromo-2,4-dichloro-5-nitrobenzene
Formel: C₆H₂BrCl₂NO₂
Molecular Weight: 270,9000 g/mol
MDL: MFCD16038032
CAS-Nummer: 65001-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR