2-Brom-6-nitrobenzoesäure

Lieferant: COMBI-BLOCKS INC.

COBBSS-3792-1GEA 187 EUR
COBBSS-3792-1G
2-Brom-6-nitrobenzoesäure
2-Bromo-6-nitrobenzoic acid
Formel: C₇H₄BrNO₄
Molecular Weight: 246,0200 g/mol
MDL: MFCD13688997
CAS-Nummer: 38876-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR