3-Brom-o-anisaldehyd
Lieferant: COMBI-BLOCKS INC.
COBBCA-4234-5GEA
521
EUR
COBBCA-4234-5G
COBBCA-4234-1G
3-Brom-o-anisaldehyd
3-Brom-o-anisaldehyd
Formel:
C₈H₇BrO₂ Molecular Weight: 215,0400 g/mol |
MDL:
MFCD09834811 CAS-Nummer: 88275-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...