4-Brom-N,N-diethylbenzolsulfonamid
Lieferant: COMBI-BLOCKS INC.
COBBOT-0087-5GEA
220
EUR
COBBOT-0087-5G
COBBOT-0087-1G
4-Brom-N,N-diethylbenzolsulfonamid
4-Brom-N,N-diethylbenzolsulfonamid
Formel:
C₁₀H₁₄BrNO₂S Molecular Weight: 292,1900 g/mol |
MDL:
MFCD00587586 CAS-Nummer: 90944-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...