4-Ethylbenzamid ≥98%
Lieferant: Thermo Scientific
L08339.06EA
63
EUR
L08339.06
L08339.14
4-Ethylbenzamid ≥98%
4-Ethylbenzamid
Formel:
C₉H₁₁NO Molecular Weight: 149,19 g/mol Schmelzpunkt: 160…166 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017616 CAS-Nummer: 33695-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...